Product Details

SNP ID
rs4955811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179018054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACTCAGGTACCAAAAGAGGGCA[A/G]AACAATGACCAGCCACTCCAAAAGA
Phenotype
MIM: 606452
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZMAT3 PubMed Links

Gene Details

Gene
ZMAT3
Gene Name
zinc finger matrin-type 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022470.3 8164 UTR 3 NP_071915.1
NM_152240.2 8164 UTR 3 NP_689426.1
XM_005247706.2 8164 UTR 3 XP_005247763.1
XM_011513073.1 8164 UTR 3 XP_011511375.1

View Full Product Details