Product Details
- SNP ID
-
rs7724147
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:95466326 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAACAATTTTTTTAAAGATAATAC[A/G]AACATTCCATTAGGATTCTGTTACC
- Phenotype
-
MIM: 614589
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TTC37
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143586581] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TTC37
- Gene Name
- tetratricopeptide repeat domain 37
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014639.3 |
|
Intron |
|
|
NP_055454.1 |
View Full Product Details