Product Details

SNP ID
rs8043607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:8795239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCAAAGCAAGAGCCTCTAGACCT[C/G]TGGGTGGTGAACAAATCTCAACTGG
Phenotype
MIM: 601785
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PMM2 PubMed Links

Gene Details

Gene
PMM2
Gene Name
phosphomannomutase 2
There are no transcripts associated with this gene.

Gene
TMEM186
Gene Name
transmembrane protein 186
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015421.3 1389 UTR 3 NP_056236.2

View Full Product Details