Product Details

SNP ID
rs6760556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:196134586 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGATCGCTCCACTAGAGAACAG[A/T]TAACGGCTTACTTCCTTTAAAGTGC
Phenotype
MIM: 604727
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
STK17B PubMed Links

Gene Details

Gene
STK17B
Gene Name
serine/threonine kinase 17b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004226.3 4304 UTR 3 NP_004217.1
XM_011512168.2 4304 UTR 3 XP_011510470.1
XM_011512169.1 4304 UTR 3 XP_011510471.1
XM_011512170.1 4304 UTR 3 XP_011510472.1
XM_011512171.2 4304 UTR 3 XP_011510473.1

View Full Product Details