Product Details

SNP ID
rs7167569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64696718 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTACTAACATATCACATCCTGCTGC[C/T]TCTAATTTAAAGTGTCTGTGAAACT
Phenotype
MIM: 604152
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OAZ2 PubMed Links

Gene Details

Gene
OAZ2
Gene Name
ornithine decarboxylase antizyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301302.1 Intron NP_001288231.1
NM_002537.3 Intron NP_002528.1

View Full Product Details