Product Details

SNP ID
rs6660150
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:205086959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTACTAATCTCAACCACTGGT[C/T]TAACTCATGACAGTCTCAAAATGAA
Phenotype
MIM: 190197 MIM: 600697
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNTN2 PubMed Links

Gene Details

Gene
CNTN2
Gene Name
contactin 2
There are no transcripts associated with this gene.

Gene
RBBP5
Gene Name
RB binding protein 5, histone lysine methyltransferase complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193272.1 3589 UTR 3 NP_001180201.1
NM_001193273.1 3589 UTR 3 NP_001180202.1
NM_005057.3 3589 UTR 3 NP_005048.2
XM_011509833.2 3589 UTR 3 XP_011508135.1
Gene
TMEM81
Gene Name
transmembrane protein 81
There are no transcripts associated with this gene.

View Full Product Details