Product Details

SNP ID
rs7591664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:172024347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAAAAGGGGAAGGGTGTGAAAG[C/T]AGCCATTTAGATAAACAGAAAAATG
Phenotype
MIM: 610267
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
METAP1D PubMed Links

Gene Details

Gene
METAP1D
Gene Name
methionyl aminopeptidase type 1D (mitochondrial)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322278.1 Intron NP_001309207.1
NM_001322279.1 Intron NP_001309208.1
NM_199227.2 Intron NP_954697.1
XM_017003750.1 Intron XP_016859239.1

View Full Product Details