Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001098272.2 | 5094 | UTR 3 | NP_001091742.1 | ||
NM_001324219.1 | 5094 | UTR 3 | NP_001311148.1 | ||
NM_001324220.1 | 5094 | UTR 3 | NP_001311149.1 | ||
NM_001324222.1 | 5094 | UTR 3 | NP_001311151.1 | ||
NM_001324223.1 | 5094 | UTR 3 | NP_001311152.1 | ||
NM_001324224.1 | 5094 | UTR 3 | NP_001311153.1 | ||
NM_002130.7 | 5094 | UTR 3 | NP_002121.4 | ||
XM_011514036.2 | 5094 | Intron | XP_011512338.1 | ||
XM_017009412.1 | 5094 | Intron | XP_016864901.1 |