Product Details

SNP ID
rs6984494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:16108866 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGCTTTCATTTGCCACTATTAAA[A/G]AAAGAACAGTAGAGTAATTCCATAA
Phenotype
MIM: 153622
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSR1 PubMed Links

Gene Details

Gene
MSR1
Gene Name
macrophage scavenger receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002445.3 2773 Intron NP_002436.1
NM_138715.2 2773 UTR 3 NP_619729.1
NM_138716.2 2773 UTR 3 NP_619730.1

View Full Product Details