Product Details

SNP ID
rs7918266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88179809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATTCTTAGGTAGAAATCAAAGT[A/G]TTTGCTATGTCTTTTAATTATTAAA
Phenotype
MIM: 609360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNLS PubMed Links

Gene Details

Gene
RNLS
Gene Name
renalase, FAD dependent amine oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031709.2 4793 Intron NP_001026879.2
NM_018363.3 4793 Intron NP_060833.1
XM_005269946.1 4793 Intron XP_005270003.1
XM_005269947.1 4793 Intron XP_005270004.1
XM_005269948.2 4793 Intron XP_005270005.1
XM_005269949.4 4793 Intron XP_005270006.1
XM_005269950.3 4793 Intron XP_005270007.1
XM_011539924.2 4793 UTR 3 XP_011538226.1
XM_011539927.2 4793 Intron XP_011538229.1
XM_017016380.1 4793 Intron XP_016871869.1
XM_017016381.1 4793 Intron XP_016871870.1
XM_017016382.1 4793 UTR 3 XP_016871871.1
XM_017016383.1 4793 Intron XP_016871872.1
XM_017016384.1 4793 Intron XP_016871873.1
XM_017016385.1 4793 Intron XP_016871874.1

View Full Product Details