Product Details

SNP ID
rs7711180
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:53567666 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCTGCTATGCTTTCTGAGTTCTC[C/G]TGTCTGCTTTTTAATAGTGTTATAT
Phenotype
MIM: 602694
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
NDUFS4 PubMed Links

Gene Details

Gene
NDUFS4
Gene Name
NADH:ubiquinone oxidoreductase subunit S4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318051.1 Intron NP_001304980.1
NM_002495.3 Intron NP_002486.1
XM_017009491.1 Intron XP_016864980.1

View Full Product Details