Product Details

SNP ID
rs6886768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:74631945 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTACACGTCAAGCACTGGGCTAG[C/G]TATTTTCGCCTCTTTTAACTCCCCT
Phenotype
MIM: 605173 MIM: 606873
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ENC1 PubMed Links

Gene Details

Gene
ENC1
Gene Name
ectodermal-neural cortex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256574.1 Intron NP_001243503.1
NM_001256575.1 Intron NP_001243504.1
NM_001256576.1 Intron NP_001243505.1
NM_003633.3 Intron NP_003624.1
XM_011543696.2 Intron XP_011541998.1
XM_011543697.2 Intron XP_011541999.1
Gene
HEXB
Gene Name
hexosaminidase subunit beta
There are no transcripts associated with this gene.

View Full Product Details