Product Details
- SNP ID
-
rs7720633
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:73630099 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACATGATCATTGTGCCTTTAAAAA[A/C]TATACACTTGAAATGTTAATATGCT
- Phenotype
-
MIM: 612790
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF28
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115253433] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGEF28
- Gene Name
- Rho guanine nucleotide exchange factor 28
View Full Product Details