Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001135745.1 | Intron | NP_001129217.1 | |||
NM_020919.3 | Intron | NP_065970.2 | |||
XM_006712654.2 | Intron | XP_006712717.1 | |||
XM_006712655.3 | Intron | XP_006712718.1 | |||
XM_011511530.2 | Intron | XP_011509832.1 | |||
XM_017004569.1 | Intron | XP_016860058.1 | |||
XM_017004570.1 | Intron | XP_016860059.1 | |||
XM_017004571.1 | Intron | XP_016860060.1 | |||
XM_017004572.1 | Intron | XP_016860061.1 |