Product Details

SNP ID
rs6808125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:28349263 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGAGGTCCCCCTTCAGGGGCGCCCT[C/T]TGGTGCGTCTTTTTCACTCAGTGTC
Phenotype
MIM: 609916
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AZI2 PubMed Links

Gene Details

Gene
AZI2
Gene Name
5-azacytidine induced 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134432.1 Intron NP_001127904.1
NM_001134433.1 Intron NP_001127905.1
NM_001271650.1 Intron NP_001258579.1
NM_022461.4 Intron NP_071906.1
XM_005265392.2 Intron XP_005265449.1
XM_011534009.2 Intron XP_011532311.1
XM_011534010.2 Intron XP_011532312.1
XM_011534011.2 Intron XP_011532313.1
Gene
ZCWPW2
Gene Name
zinc finger CW-type and PWWP domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040432.3 Intron NP_001035522.1
NM_001324169.1 Intron NP_001311098.1
NM_001324170.1 Intron NP_001311099.1
XM_017005755.1 Intron XP_016861244.1
XM_017005756.1 Intron XP_016861245.1
XM_017005757.1 Intron XP_016861246.1
XM_017005758.1 Intron XP_016861247.1
XM_017005759.1 Intron XP_016861248.1

View Full Product Details