Product Details

SNP ID
rs7873152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:15555460 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCCATGGCTTGTCAGTTTCGATA[C/T]GGGGAAAAATGATATTTTTAAGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC171 PubMed Links

Gene Details

Gene
CCDC171
Gene Name
coiled-coil domain containing 171
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173550.2 Intron NP_775821.2
XM_005251397.2 Intron XP_005251454.1
XM_005251398.3 Intron XP_005251455.1
XM_005251399.4 Intron XP_005251456.1
XM_011517788.2 Intron XP_011516090.1
XM_011517791.2 Intron XP_011516093.1
XM_011517796.2 Intron XP_011516098.1
XM_017014431.1 Intron XP_016869920.1
XM_017014432.1 Intron XP_016869921.1
XM_017014433.1 Intron XP_016869922.1
XM_017014434.1 Intron XP_016869923.1
XM_017014435.1 Intron XP_016869924.1
XM_017014436.1 Intron XP_016869925.1
XM_017014437.1 Intron XP_016869926.1
XM_017014438.1 Intron XP_016869927.1
XM_017014439.1 Intron XP_016869928.1
XM_017014440.1 Intron XP_016869929.1
XM_017014441.1 Intron XP_016869930.1
XM_017014442.1 Intron XP_016869931.1
XM_017014443.1 Intron XP_016869932.1
XM_017014444.1 Intron XP_016869933.1

View Full Product Details