Product Details

SNP ID
rs6896186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157138964 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATCATTTCAATTACTCGTTCCAG[A/G]TGCTTTTGAAATCTCTCAGCTGTTT
Phenotype
MIM: 605045
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED7 PubMed Links

Gene Details

Gene
MED7
Gene Name
mediator complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100816.1 576 Silent Mutation CAC,CAT H156H NP_001094286.1
NM_004270.4 576 Silent Mutation CAC,CAT H156H NP_004261.1

View Full Product Details