Product Details

SNP ID
rs6954722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:152137070 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTTATTGAGGTTCCATGGGACCC[C/T]GGGGGAACCCTGAAAAGAGGCATTG
Phenotype
MIM: 606833
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KMT2C PubMed Links

Gene Details

Gene
KMT2C
Gene Name
lysine methyltransferase 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170606.2 Intron NP_733751.2
XM_005250025.4 Intron XP_005250082.1
XM_005250026.3 Intron XP_005250083.1
XM_005250027.4 Intron XP_005250084.1
XM_005250028.4 Intron XP_005250085.1
XM_005250031.4 Intron XP_005250088.1
XM_006716077.3 Intron XP_006716140.1
XM_006716078.3 Intron XP_006716141.1
XM_006716079.3 Intron XP_006716142.1
XM_011516450.2 Intron XP_011514752.1
XM_011516451.2 Intron XP_011514753.1
XM_011516452.2 Intron XP_011514754.1
XM_011516453.2 Intron XP_011514755.1
XM_011516454.2 Intron XP_011514756.1
XM_011516456.2 Intron XP_011514758.1
XM_017012480.1 Intron XP_016867969.1
XM_017012481.1 Intron XP_016867970.1
XM_017012482.1 Intron XP_016867971.1
XM_017012483.1 Intron XP_016867972.1
XM_017012484.1 Intron XP_016867973.1
XM_017012485.1 Intron XP_016867974.1
XM_017012486.1 Intron XP_016867975.1
XM_017012487.1 Intron XP_016867976.1
XM_017012488.1 Intron XP_016867977.1
XM_017012489.1 Intron XP_016867978.1
XM_017012490.1 Intron XP_016867979.1
Gene
LOC731075
Gene Name
uncharacterized LOC731075
There are no transcripts associated with this gene.

Gene
YBX1P4
Gene Name
Y-box binding protein 1 pseudogene 4
There are no transcripts associated with this gene.

View Full Product Details