Product Details
- SNP ID
-
rs6867419
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:80321119 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAATAATGAAGTGTCGGCTAAGCAT[A/G]AGCTGGAGGAACAGGTGAAGAAACT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SPZ1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35337118] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPZ1
- Gene Name
- spermatogenic leucine zipper 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032567.3 |
1149 |
Missense Mutation |
AAG,GAG |
K302E |
NP_115956.3 |
View Full Product Details