Product Details
- SNP ID
-
rs7187298
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:81989176 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTCTTAGGCCAACTAGGACATATG[C/G]TTATCATCTTTATATACCCGGTGAC
- Phenotype
-
MIM: 616164
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SDR42E1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113509841] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SDR42E1
- Gene Name
- short chain dehydrogenase/reductase family 42E, member 1
View Full Product Details