Product Details

SNP ID
rs6722185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218660859 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGAGGGTATTGACCCACACAGTA[A/G]TGCTGACCTGTGGCCAGGGCTGGGG
Phenotype
MIM: 603647 MIM: 616014 MIM: 604083
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCS1L PubMed Links

Gene Details

Gene
BCS1L
Gene Name
BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079866.1 908 Intron NP_001073335.1
NM_001257342.1 908 Intron NP_001244271.1
NM_001257343.1 908 Intron NP_001244272.1
NM_001257344.1 908 Intron NP_001244273.1
NM_001318836.1 908 Intron NP_001305765.1
NM_001320717.1 908 Intron NP_001307646.1
NM_004328.4 908 Intron NP_004319.1
XM_005246748.2 908 Intron XP_005246805.1
XM_005246749.3 908 Intron XP_005246806.1
XM_006712678.1 908 Intron XP_006712741.1
XM_017004631.1 908 Intron XP_016860120.1
XM_017004632.1 908 Intron XP_016860121.1
XM_017004633.1 908 UTR 5 XP_016860122.1
XM_017004634.1 908 Intron XP_016860123.1
Gene
RNF25
Gene Name
ring finger protein 25
There are no transcripts associated with this gene.

Gene
ZNF142
Gene Name
zinc finger protein 142
There are no transcripts associated with this gene.

View Full Product Details