Product Details

SNP ID
rs7927324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9780445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCATCAGAGATACAACTCTGGAT[C/T]TTGTCCATCCATTGCTGGGCACTCT
Phenotype
MIM: 607697
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SBF2 PubMed Links

Gene Details

Gene
SBF2
Gene Name
SET binding factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030962.3 5685 Silent Mutation AAA,AAG K1841K NP_112224.1
XM_005253154.4 5685 Silent Mutation AAA,AAG K1873K XP_005253211.1
XM_005253155.4 5685 Silent Mutation AAA,AAG K1830K XP_005253212.1
XM_011520394.2 5685 Silent Mutation AAA,AAG K1835K XP_011518696.1
XM_011520395.2 5685 Intron XP_011518697.1
XM_011520396.2 5685 Intron XP_011518698.1
XM_017018372.1 5685 Silent Mutation AAA,AAG K1827K XP_016873861.1
XM_017018373.1 5685 Silent Mutation AAA,AAG K1827K XP_016873862.1
XM_017018374.1 5685 Silent Mutation AAA,AAG K1798K XP_016873863.1
XM_017018375.1 5685 Silent Mutation AAA,AAG K1794K XP_016873864.1
XM_017018376.1 5685 Intron XP_016873865.1
XM_017018377.1 5685 Intron XP_016873866.1
Gene
SBF2-AS1
Gene Name
SBF2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details