Product Details
- SNP ID
-
rs6966356
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:4858129 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGATGTGAGAAAAAGAAATACTGT[A/G]TACTAGATATCAGAAAGTAATGGTG
- Phenotype
-
MIM: 607436
MIM: 611491
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PAPOLB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77021497] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PAPOLB
- Gene Name
- poly(A) polymerase beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020144.4 |
3866 |
UTR 3 |
|
|
NP_064529.4 |
- Gene
- RADIL
- Gene Name
- Rap associating with DIL domain
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018059.4 |
3866 |
Intron |
|
|
NP_060529.4 |
View Full Product Details