Product Details

SNP ID
rs7294618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50119192 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGACACTATTTCAGTTATATTAT[A/G]GCTTAAACAAGCTTTAGTGTTGACT
Phenotype
MIM: 614478 MIM: 138420
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX14 PubMed Links

Gene Details

Gene
COX14
Gene Name
COX14, cytochrome c oxidase assembly factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257133.1 Intron NP_001244062.1
NM_001257134.1 Intron NP_001244063.1
NM_032901.3 Intron NP_116290.1
Gene
GPD1
Gene Name
glycerol-3-phosphate dehydrogenase 1
There are no transcripts associated with this gene.

View Full Product Details