Product Details

SNP ID
rs7565273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:114444460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTCTGAGGTCACAGGAATCAAG[C/T]ATTTATTTTCCCTTTTCTTTATTGA
Phenotype
MIM: 608209
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPP10 PubMed Links

Gene Details

Gene
DPP10
Gene Name
dipeptidyl peptidase like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004360.3 Intron NP_001004360.2
NM_001178034.1 Intron NP_001171505.1
NM_001178036.1 Intron NP_001171507.1
NM_001178037.1 Intron NP_001171508.1
NM_001321905.1 Intron NP_001308834.1
NM_001321906.1 Intron NP_001308835.1
NM_001321907.1 Intron NP_001308836.1
NM_001321908.1 Intron NP_001308837.1
NM_001321909.1 Intron NP_001308838.1
NM_001321910.1 Intron NP_001308839.1
NM_001321911.1 Intron NP_001308840.1
NM_001321912.1 Intron NP_001308841.1
NM_001321913.1 Intron NP_001308842.1
NM_001321914.1 Intron NP_001308843.1
NM_020868.4 Intron NP_065919.2
XM_017004565.1 Intron XP_016860054.1
XM_017004566.1 Intron XP_016860055.1

View Full Product Details