Product Details

SNP ID
rs7237253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:49783539 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAACATTTATTTGATTTCCTTATA[A/G]TCATATTATCTAAGGCAGAAGTGGT
Phenotype
MIM: 604770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAA2 PubMed Links

Gene Details

Gene
ACAA2
Gene Name
acetyl-CoA acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006111.2 1899 UTR 3 NP_006102.2

View Full Product Details