Product Details

SNP ID
rs7067848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49131233 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACAGATTTGTAATAGTGAAAAAA[A/G]TAGGGCTGGAGAGTAATTTGCAGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM170B PubMed Links

Gene Details

Gene
FAM170B
Gene Name
family with sequence similarity 170 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164484.1 1334 UTR 3 NP_001157956.1
Gene
FAM170B-AS1
Gene Name
FAM170B antisense RNA 1
There are no transcripts associated with this gene.

Gene
VSTM4
Gene Name
V-set and transmembrane domain containing 4
There are no transcripts associated with this gene.

View Full Product Details