Product Details
- SNP ID
-
rs6424261
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
49
- Location
-
Chr.1:232954477 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATGCTAGGAGTTTTCTCTCTCTTT[C/T]CATTGTTGGGTAGTCATGATGCCAC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NTPCR
PubMed Links
Gene Details
- Gene
- NTPCR
- Gene Name
- nucleoside-triphosphatase, cancer-related
There are no transcripts associated with this gene.
View Full Product Details