Product Details

SNP ID
rs7677151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:36011594 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTTAGAGATAGTGACTGTGATGA[C/T]TAGTCACTTCTGGCAGGAAGGGAAT
Phenotype
MIM: 606645
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARAP2 PubMed Links

Gene Details

Gene
ARAP2
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015230.3 Intron NP_056045.2
XM_017007698.1 Intron XP_016863187.1
XM_017007699.1 Intron XP_016863188.1
XM_017007700.1 Intron XP_016863189.1
XM_017007701.1 Intron XP_016863190.1
XM_017007702.1 Intron XP_016863191.1

View Full Product Details