Product Details

SNP ID
rs5933549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53161457 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAGATATTACTGATACATCAGCT[C/T]TCGTATTCATAAGCCCATAAAATTT
Phenotype
MIM: 314690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM5C PubMed Links

Gene Details

Gene
KDM5C
Gene Name
lysine demethylase 5C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146702.1 Intron NP_001140174.1
NM_001282622.1 Intron NP_001269551.1
NM_004187.3 Intron NP_004178.2
XM_005262035.4 Intron XP_005262092.1
XM_011530824.2 Intron XP_011529126.1
XM_011530825.2 Intron XP_011529127.1
XM_011530826.2 Intron XP_011529128.1
XM_011530827.2 Intron XP_011529129.1
XM_011530828.1 Intron XP_011529130.1
XM_011530829.2 Intron XP_011529131.1
XM_011530830.2 Intron XP_011529132.1
XM_011530831.2 Intron XP_011529133.1
XM_017029886.1 Intron XP_016885375.1
XM_017029887.1 Intron XP_016885376.1
XM_017029888.1 Intron XP_016885377.1
XM_017029889.1 Intron XP_016885378.1

View Full Product Details