Product Details

SNP ID
rs7184719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:10747957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTCACATTTTTGAAATGTTTTT[C/T]GTTTGTTTGAGACAGGATCTCACTC
Phenotype
MIM: 600280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUBP1 PubMed Links

Gene Details

Gene
NUBP1
Gene Name
nucleotide binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278506.1 Intron NP_001265435.1
NM_001323594.1 Intron NP_001310523.1
NM_001323595.1 Intron NP_001310524.1
NM_001323596.1 Intron NP_001310525.1
NM_001323597.1 Intron NP_001310526.1
NM_002484.3 Intron NP_002475.2
XM_017023252.1 Intron XP_016878741.1
Gene
TEKT5
Gene Name
tektin 5
There are no transcripts associated with this gene.

View Full Product Details