Product Details

SNP ID
rs6998333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:29334572 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGTCCCTGTGGTGGGAATCCAGT[C/T]TGGGAAAGCAGGACTGTTTTAGCAA
Phenotype
MIM: 602747
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP4 PubMed Links

Gene Details

Gene
DUSP4
Gene Name
dual specificity phosphatase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001394.6 6671 UTR 3 NP_001385.1
NM_057158.3 6671 UTR 3 NP_476499.1
XM_011544428.2 6671 UTR 3 XP_011542730.1

View Full Product Details