Product Details
- SNP ID
-
rs7716437
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:7401153 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAAGCTGATGTCCTCAATTAGTAC[A/C]TGCAAAGCCCTTAGAACAATGCCTG
- Phenotype
-
MIM: 103071
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADCY2
PubMed Links
Gene Details
- Gene
- ADCY2
- Gene Name
- adenylate cyclase 2
View Full Product Details