Product Details

SNP ID
rs7259127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54471021 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTCGCTTCACTTCTGCCAGCCTG[C/G]GTGTCCTCTTAAATAGAAGCCTCTG
Phenotype
MIM: 609171 MIM: 616575
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDC42EP5 PubMed Links

Gene Details

Gene
CDC42EP5
Gene Name
CDC42 effector protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145057.3 Intron NP_659494.2
Gene
LENG8
Gene Name
leukocyte receptor cluster member 8
There are no transcripts associated with this gene.

Gene
LENG9
Gene Name
leukocyte receptor cluster member 9
There are no transcripts associated with this gene.

View Full Product Details