Product Details

SNP ID
rs7824139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:21909544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGTCAGGTCGAGGGGGCAGGG[G/T]CTCCTCAATGCTGTCGTACAGAGGG
Phenotype
MIM: 604997
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DOK2 PubMed Links

Gene Details

Gene
DOK2
Gene Name
docking protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317800.1 1025 Missense Mutation ACC,CCC T182P NP_001304729.1
NM_003974.3 1025 Missense Mutation ACC,CCC T336P NP_003965.2
NM_201349.2 1025 Missense Mutation ACC,CCC T182P NP_958728.1
XM_005273680.4 1025 Missense Mutation ACC,CCC T242P XP_005273737.1

View Full Product Details