Product Details

SNP ID
rs8104220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55401992 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCCCAAAAGGACACGATGCCATA[C/G]GCCCAATGGTTTAAAATGTCCCTGG
Phenotype
MIM: 603638 MIM: 610309
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RPL28 PubMed Links

Gene Details

Gene
RPL28
Gene Name
ribosomal protein L28
There are no transcripts associated with this gene.

Gene
UBE2S
Gene Name
ubiquitin conjugating enzyme E2 S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014501.2 Intron NP_055316.2
XM_011526752.1 Intron XP_011525054.1

View Full Product Details