Product Details

SNP ID
rs7086629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71969470 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATCTCTGCCCTGAGAGCCCCCAG[C/T]ACAGATGTCCATAGAACGGGAAAGG
Phenotype
MIM: 603799
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST3 PubMed Links

Gene Details

Gene
CHST3
Gene Name
carbohydrate sulfotransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004273.4 1015 Intron NP_004264.2
XM_006718075.3 1015 Intron XP_006718138.1
XM_011540369.2 1015 UTR 5 XP_011538671.1

View Full Product Details