Product Details

SNP ID
rs7093449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:23712401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCAAGATGTTCATTGACTTGGC[G/T]ACCATTATAAGCCAAGTCAATGAAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1217 PubMed Links

Gene Details

Gene
KIAA1217
Gene Name
KIAA1217
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098500.2 Intron NP_001091970.1
NM_001282767.1 Intron NP_001269696.1
NM_001282768.1 Intron NP_001269697.1
NM_001282769.1 Intron NP_001269698.1
NM_001282770.1 Intron NP_001269699.1
NM_001321681.1 Intron NP_001308610.1
NM_019590.4 Intron NP_062536.2
XM_005252516.3 Intron XP_005252573.1
XM_011519552.2 Intron XP_011517854.2
XM_011519555.2 Intron XP_011517857.2
XM_011519558.2 Intron XP_011517860.2
XM_011519559.2 Intron XP_011517861.2
XM_011519562.2 Intron XP_011517864.2
XM_011519564.2 Intron XP_011517866.1
XM_011519565.2 Intron XP_011517867.2
XM_011519566.2 Intron XP_011517868.2
XM_017016416.1 Intron XP_016871905.1
XM_017016417.1 Intron XP_016871906.1
XM_017016418.1 Intron XP_016871907.1
XM_017016419.1 Intron XP_016871908.1
XM_017016420.1 Intron XP_016871909.1
XM_017016421.1 Intron XP_016871910.1
XM_017016422.1 Intron XP_016871911.1
XM_017016423.1 Intron XP_016871912.1
XM_017016424.1 Intron XP_016871913.1
XM_017016425.1 Intron XP_016871914.1
XM_017016426.1 Intron XP_016871915.1
XM_017016427.1 Intron XP_016871916.1
XM_017016428.1 Intron XP_016871917.1
XM_017016429.1 Intron XP_016871918.1

View Full Product Details