Product Details

SNP ID
rs7908746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:3138537 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCACGCTGACCCCTACCCACGCC[C/T]GCTTCTGCAGCAGGGATGCAGTCAG
Phenotype
MIM: 171840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PFKP PubMed Links

Gene Details

Gene
PFKP
Gene Name
phosphofructokinase, platelet
There are no transcripts associated with this gene.

Gene
PITRM1
Gene Name
pitrilysin metallopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242307.1 Intron NP_001229236.1
NM_001242309.1 Intron NP_001229238.1
NM_014889.3 Intron NP_055704.2
XM_005252345.3 Intron XP_005252402.1
XM_017015470.1 Intron XP_016870959.1
XM_017015471.1 Intron XP_016870960.1
XM_017015472.1 Intron XP_016870961.1
XM_017015473.1 Intron XP_016870962.1
Gene
PITRM1-AS1
Gene Name
PITRM1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SNORD142
Gene Name
small nucleolar RNA, C/D box 142
There are no transcripts associated with this gene.

View Full Product Details