Product Details

SNP ID
rs7202756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69362555 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGAGAGCCACAAGGGAGTTATAC[C/T]ATCAAAGATTAGAGTAAAAAGCAAA
Phenotype
MIM: 602027
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TERF2 PubMed Links

Gene Details

Gene
TERF2
Gene Name
telomeric repeat binding factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005652.4 Intron NP_005643.2
XM_005256121.3 Intron XP_005256178.1
XM_005256122.3 Intron XP_005256179.1
XM_005256123.3 Intron XP_005256180.1
XM_005256124.3 Intron XP_005256181.1

View Full Product Details