Product Details

SNP ID
rs6947138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6346759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATCACCTGTGTCAGGCACCAGC[C/T]ACCTGAAGACCTGCTACTGGGACAA
Phenotype
MIM: 616628
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM220A PubMed Links

Gene Details

Gene
FAM220A
Gene Name
family with sequence similarity 220 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037163.1 Intron NP_001032240.1

View Full Product Details