Product Details

SNP ID
rs6580873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52288141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTGAGATGTGCGACTGGAGAATG[A/C]GGATCTCCTGCAGGAGGTGAGGGCA
Phenotype
MIM: 602153 MIM: 601928
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRT81 PubMed Links

Gene Details

Gene
KRT81
Gene Name
keratin 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002281.3 793 Missense Mutation NP_002272.2
Gene
KRT86
Gene Name
keratin 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320198.1 793 Intron NP_001307127.1
XM_005268866.4 793 Intron XP_005268923.1
XM_017019296.1 793 Intron XP_016874785.1

View Full Product Details