Product Details

SNP ID
rs7722605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154049066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATATCTTCATACTTGAATTACTC[A/C]AATTTTGAGCTCCATGGTTATGATT
Phenotype
MIM: 600491
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MFAP3 PubMed Links

Gene Details

Gene
MFAP3
Gene Name
microfibrillar associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135037.1 Intron NP_001128509.1
NM_001242336.1 Intron NP_001229265.1
NM_005927.4 Intron NP_005918.1

View Full Product Details