Product Details

SNP ID
rs6442020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46694155 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTGGGTTCGTGGAACTTGGAGCC[G/T]GTAGAGGGGTATACTGAGGCGCCCT
Phenotype
MIM: 612402 MIM: 607237
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALS2CL PubMed Links

Gene Details

Gene
ALS2CL
Gene Name
ALS2 C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190707.1 356 Intron NP_001177636.1
NM_147129.4 356 Intron NP_667340.2
XM_005265025.1 356 Intron XP_005265082.1
XM_006713091.2 356 Intron XP_006713154.1
XM_006713093.3 356 Intron XP_006713156.1
XM_006713094.3 356 Intron XP_006713157.1
XM_011533572.2 356 Intron XP_011531874.1
XM_017006120.1 356 Intron XP_016861609.1
Gene
TMIE
Gene Name
transmembrane inner ear
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147196.2 356 Intron NP_671729.2
XM_006713097.3 356 UTR 5 XP_006713160.1

View Full Product Details