Product Details

SNP ID
rs6442051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46981892 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCCCCTCAGGAGGTGGTTATGGC[G/T]CATATCTGTGCTGAGCCTCCCCCTC
Phenotype
MIM: 614169
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC12 PubMed Links

Gene Details

Gene
CCDC12
Gene Name
coiled-coil domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277074.1 Intron NP_001264003.1
NM_144716.5 Intron NP_653317.2
XM_011533390.2 Intron XP_011531692.1
XM_011533391.2 Intron XP_011531693.1
XM_011533392.2 Intron XP_011531694.1
XM_011533393.2 Intron XP_011531695.1
XM_011533394.2 Intron XP_011531696.1
XM_011533396.2 Intron XP_011531698.1
Gene
NBEAL2
Gene Name
neurobeachin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015175.2 Intron NP_055990.1
XM_006713072.3 Intron XP_006713135.1
XM_011533533.2 Intron XP_011531835.1
XM_017006010.1 Intron XP_016861499.1
XM_017006011.1 Intron XP_016861500.1
XM_017006012.1 Intron XP_016861501.1
XM_017006013.1 Intron XP_016861502.1
XM_017006014.1 Intron XP_016861503.1
XM_017006015.1 Intron XP_016861504.1
XM_017006016.1 Intron XP_016861505.1
XM_017006017.1 Intron XP_016861506.1

View Full Product Details