Product Details

SNP ID
rs7484157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119015887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGGAGCATCTCCACCCTTGGT[A/T]TTTCTGGTGTAAATTACTTGAGCTC
Phenotype
MIM: 180465 MIM: 602671 MIM: 610971
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC84 PubMed Links

Gene Details

Gene
CCDC84
Gene Name
coiled-coil domain containing 84
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198489.2 Intron NP_940891.1
XM_011542799.1 Intron XP_011541101.1
XM_011542800.2 Intron XP_011541102.1
XM_017017652.1 Intron XP_016873141.1
Gene
MIR3656
Gene Name
microRNA 3656
There are no transcripts associated with this gene.

Gene
RPS25
Gene Name
ribosomal protein S25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001028.2 Intron NP_001019.1
Gene
SLC37A4
Gene Name
solute carrier family 37 member 4
There are no transcripts associated with this gene.

Gene
TRAPPC4
Gene Name
trafficking protein particle complex 4
There are no transcripts associated with this gene.

View Full Product Details