Product Details

SNP ID
rs7125935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:43264275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCATCCTGCTTTCCCTGTCTCT[C/T]AAATGGGAGATGCCTATCCTTACCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HNRNPKP3 PubMed Links

Gene Details

Gene
HNRNPKP3
Gene Name
heterogeneous nuclear ribonucleoprotein K pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details