Product Details

SNP ID
rs8116767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44357658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGTCATACTCATTATATAGCA[A/G]ACATTCAGCAATTATTAGAATGGTT
Phenotype
MIM: 600281
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HNF4A PubMed Links

Gene Details

Gene
HNF4A
Gene Name
hepatocyte nuclear factor 4 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000457.4 Intron NP_000448.3
NM_001030003.2 Intron NP_001025174.1
NM_001030004.2 Intron NP_001025175.1
NM_001258355.1 Intron NP_001245284.1
NM_001287182.1 Intron NP_001274111.1
NM_001287183.1 Intron NP_001274112.1
NM_001287184.1 Intron NP_001274113.1
NM_175914.4 Intron NP_787110.2
NM_178849.2 Intron NP_849180.1
NM_178850.2 Intron NP_849181.1
XM_005260407.3 Intron XP_005260464.1
Gene
R3HDML
Gene Name
R3H domain containing like
There are no transcripts associated with this gene.

View Full Product Details