Product Details

SNP ID
rs7353566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45479645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAACCCTATGTTGTAATTAGCAC[G/T]GTTCCACTGGCACCTAAAGACACGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WFDC2 PubMed Links

Gene Details

Gene
WFDC2
Gene Name
WAP four-disulfide core domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006103.3 Intron NP_006094.3

View Full Product Details