Product Details

SNP ID
rs6791196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:130347861 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATCCAGTACACACTGAGCAAGTG[G/T]AAAGTGCAAAGGAAGAGGAGTTGGG
Phenotype
MIM: 611916
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COL6A5 PubMed Links

Gene Details

Gene
COL6A5
Gene Name
collagen type VI alpha 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278298.1 Intron NP_001265227.1
NM_153264.6 Intron NP_694996.5
XM_011512621.2 Intron XP_011510923.1
XM_011512622.2 Intron XP_011510924.1
XM_011512623.2 Intron XP_011510925.1

View Full Product Details